Learn More
ACROBiosystems Human M-CSF / CSF-1 Protein, premium grade
Supplier:  ACROBiosystems MCFH521820UG
Encompass_Preferred
Human M-CSF Protein, premium grade (MCF-H5218) is expressed from human 293 cells (HEK293). It contains AA Glu 33 - Arg 255 (Accession # P09603-1).
Catalog No. 50-233-0927