Learn More
Sino Biological Anti-MEK1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200291T40100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human MEK1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-1196