Learn More
Abcam Human MASP1 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Supplier:  Abcam AB25362410X96TESTS
Encompass_Preferred
Human MASP1 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human MASP1.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-5151