Learn More
Cusabio Technology LLC Human Mast Cell Tryptase ELISA Kit, Sandwich ELISA, 96T, Detection Range 1.56 ng/mL - 100 ng/mL, Sensitivity <0.39 ng/mL, Storage 2-8°C
Supplier: Cusabio Technology LLC CSBE09012H
This quantitative sandwich ELISA kit is designed for the precise determination of human mast cell tryptase (MCT) concentrations in biological samples. With its high sensitivity and excellent specificity, it is well-suited for researchers investigating mast cell activation, allergic reactions, and inflammatory processes.
- Target: Human mast cell tryptase (MCT)
- Application: Quantitative determination of human MCT concentrations
- Method: Sandwich ELISA
- Sample types: Serum, plasma, tissue homogenates
- Detection range: 1.56 ng/mL - 100 ng/mL
- Sensitivity: <0.39 ng/mL
- Specificity: No significant cross-reactivity observed
- Storage (unopened): 2-8°C
- Kit sizes: 96T, 5x96T, 10x96T
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.