Learn More
Sino Biological Recombinant Human c-MET/HGFR Protein (Site-Specific AF 488 Conjugation, ECD, His Tag)
Supplier:  Sino Biological 10692H86HSD100
Encompass_Preferred
A DNA sequence encoding the Human MET (NP_000236.2) (Met1-Thr932) was expressed with a polyhistidine tag at the C-terminus. The protein is site-specifically labeled with AF 488 (Excitation Max. = 495 nm, Emission Max. = 519 nm).
Catalog No. 50-299-2857
 Restricted Item