Learn More
ACROBiosystems Biotinylated Human MICA, His,Avitag (MIA-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 24 - Gln 308 (Accession # Q29983-1).
Supplier:  ACROBiosystems MIAH82E625UG
Encompass_Preferred
MHC class I polypeptide-related sequence A (MICA) belongs to the MHC class I family and MIC subfamily. MICA contains one Ig-like C1-type (immunoglobulin-like) domain. Unlike classical MHC class I molecules, MICA does not form a heterodimer with beta-2-microglobulin. MICA acts as a stress-induced self-antigen that is recognized by gamma delta T-cells. MICA is ligand for the KLRK1/NKG2D receptor. MICA bind to KLRK1 leads to cell lysis.
Catalog No. 50-225-4041