Learn More
FUJIFILM BIOSCIENCES INC Human MIP-3b (CCL19), Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 100135100UG
Encompass_Preferred
Macrophage inflammatory protein-3 beta (MIP-3 B), also called CCL19, is a chemokine that is expressed in the thymus, lymph nodes, and activated bone marrow stromal cells. MIP-3 B signals through the G protein-coupled receptor CCR7 to regulate normal lymphocyte recirculation. MIP-3 B also functions during T cell trafficking to the thymus, and in T cell and B cell homing to the lymph nodes and secondary lymphoid organs. Human MIP-3 B shows activity on mouse cells.
Catalog No. 50-900-495