Learn More
Sino Biological Recombinant Human CD10/Neprilysin Protein, Biotinylated
Supplier:  Sino Biological 10805HNCHB20
Encompass_Preferred
A DNA sequence encoding the Human MME (NP_000893.2) (Tyr52-Trp750) was expressed. The purified protein was biotinylated in vitro.
Catalog No. 50-299-2929