Learn More
Raybiotech Inc G-Series Human MMP Array 1 Kit.
Supplier:  Raybiotech Inc GSHMMP11
Encompass_Preferred
Semi-quantitative measurement of 10 human Matrix Metalloproteinase related proteins. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: MMP-1, MMP-10, MMP-13, MMP-2, MMP-3, MMP-8, MMP-9, TIMP-1, TIMP-2, TIMP-4
Catalog No. 50-195-0412