Learn More
IPHASE PHARMA SERVICES LLC Human MOCK/HEK293F Cells,8-10million
Supplier:  IPHASE PHARMA SERVICES LLC 01941A1.01
Encompass_Preferred
MOCK Cells need to be used in conjunction with with SLC transporter cells (OCT2, MATE2-K, OAT1, OAT3, OATP1B1, OATP1B3 MATE1). These products require shipment in LN2 container therefore customer need to return the container.
Catalog No. NC3966934