Learn More
Sino Biological Human MOG PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10364T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Myelin Oligodendrocyte Glycoprotein (rh Myelin Oligodendrocyte Glycoprotein; Catalog#10364-H08E; NP_996532.2; Gly30-Tyr149). Myelin Oligodendrocyte Glycoprotein specific IgG was purified by Human Myelin Oligodendrocyte Glycoprotein affinity chromatography.
Catalog No. 50-237-0487