Learn More
Sino Biological Recombinant Human CD20 Protein (Nanodisc, Full-length, Strep & His)
Supplier:  Sino Biological 11007H93HNB100
Encompass_Preferred
A DNA sequence encoding the Human MS4A1 (P11836) (Met1-Pro297) was expressed with a Strep tag at the N-terminus, a polyhistidine tag at the C-terminus. This protein was prepared using our Nanodisc platform. Nanodisc is a versatile tool for studying membrane proteins. Using styrene-maleic acid (SMA) copolymer, membrane proteins can be extracted directly from prokaryotic and eukaryotic expression systems in the absence of detergents to preserve the protein structure and function better. Compared to membrane scaffold proteins (MSPs) nanodiscs, SMA nanodiscs also have the advantage of preserving proteins' nature by maintaining native lipids surrounded without introducing any heterologous proteins, which allows studies of protein structure and functions in a native-like environment.
Catalog No. 50-299-3664