Learn More
American Research Products Inc Human MYO ELISA Kit
Supplier:  American Research Products Inc RDRMYOHU
Encompass
This Human Myoglobin (MYO) ELISA Kit is a precoated Sandwich ELISA assay designed to detect and quantify the target analyte in serum plasma tissue homogenates cell lysates cell culture supernatants and other biological fluids This product is for Research Use Only
Catalog No. 50-004-58038