Learn More
American Research Products Inc Human NACHT LRR and PYD domai
Supplier:  American Research Products Inc CSBE15885H
Encompass
Human NACHT LRR and PYD domains-containing protein 3 (NLRP3/C1orf7/CIAS1/NALP3/PYPAF1) ELISA kit from CUSABIO Detection Range 0 156 ng/ml - 10 ng/ml Sensitivity 0 039 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26819