Learn More
Sino Biological Human NBL1 Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK1016915
Encompass_Preferred
The Human NBL1 / DAND1 / DAN ELISA Pair Set is for the quantitative determination of Human NBL1 / DAND1 / DAN. This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for NBL1 / DAND1 / DAN coated on a 96-well plate. Standards and samples are added to the wells, and any NBL1 / DAND1 / DAN present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-NBL1 / DAND1 / DAN monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of NBL1 / DAND1 / DAN present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86365