Learn More
Abcam Human Nectin-4 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Supplier:  Abcam AB27035110X96TESTS
Encompass_Preferred
Human Nectin-4 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Nectin-4.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-3330