Learn More
Raybiotech Inc C-Series Human Neuro Discovery Array C2 Kit.
Supplier:  Raybiotech Inc AAHNEU28
Encompass_Preferred
Detects 30 human neurologically relevant proteins. Suitable for serum, plasma, cell culture media and other liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Adiponectin (ACRP30), BDNF, beta-NGF, CNTF, CRP (C-Reactive Protein), Eotaxin-1 (CCL11), Eotaxin-2 (MPIF-2/CCL24), Eotaxin-3 (CCL26), Fas (TNFRSF6/Apo-1), IL-4, GDNF, GM-CSF, IFN-gamma, IL-10, IL-18, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-6, IL-8 (CXCL8), LIF, MCP-1 (CCL2), MIP-1 beta (CCL4), MMP-2, MMP-3, S100 B, TARC (CCL17), TGF beta 1, TIMP-1, TNF alpha, VEGF-A
Catalog No. 50-194-8628