Learn More
American Research Products Inc Human Neurofilament protein L
Supplier:  American Research Products Inc CSBE16094H
Encompass
Human Neurofilament protein L (NF-L) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26821