Learn More
Meso Scale Discovery Human/NHP IgM Kit
SDP
Supplier:  Meso Scale Discovery K150JPD4
Encompass
The Meso Scale Discovery IGM Assay Kit is a sensitive immunoassay for quantifying IgG in samples like serum plasma and cell culture supernatant Utilizing MSD s electrochemiluminescent detection it operates on the MESO QuickPlex SQ 120 The assay offers a dynamic range of up to 4 logs delivers results in 3 hours and minimizes cross-reactivity The kit includes a 96-well plate with capture antibody detection antibody read buffer calibrator and instructions It s ideal for IgG measurement in disease research vaccine development and bioprocess applications
Catalog No. 50-000-81704
Add to Cart
Request a Quote