Learn More
Sino Biological Recombinant Human Neuregulin-4/NRG4 Protein 1
Supplier:  Sino Biological 12183HNCE1
Encompass_Preferred
A DNA sequence encoding the human NRG4 (Q8WWG1) extracellular domain (Pro 2-Phe 62) was expressed and purified, with additional two amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-224-7213