Learn More
Sino Biological Human OSMR ELISA Pair Set 5PP (Sino Biological)
Supplier:  Sino Biological SEK112265
Encompass_Preferred
Each vial contains 600 ng of recombinant OSMR. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 15,000 pg/mL is recommended.
Catalog No. 50-237-3472