Learn More
Abcam Human OX40 Antibody Pair - BSA and Azide free (TNFRSF4), 10 x 96 Tests
Supplier:  Abcam AB24407610X96TESTS
Encompass_Preferred
Human OX40 Antibody Pair - BSA and Azide free (TNFRSF4) is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human OX40 (TNFRSF4).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-5490