Learn More
ACROBiosystems ACROBiosystems Human OX40 / TNFRSF4 / CD134 Protein, Fc Tag
Supplier:  ACROBiosystems OX0H5255100UG
Encompass_Preferred
Recombinant Protein;100UG;Human OX40, Fc Tag (OX0-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accession # NP_003318).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.8 kDa. The protein migrates as 50-67 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human OX40 Ligand, His Tag (active trimer) (Cat. No. OXL-H52Q8) at 5 μg/mL (100 μL/well) can bind Human OX40, Fc Tag (Cat. No. OX0-H5255) with a linear range of 0.4-3 ng/mL (QC tested).;Quantitative Analysis of CTLA-4 x OX40 Bispecific Antibody in Human Serum by Intact Assay.
Immobilized Human OX40, Fc Tag (Cat. No. OX0-H5255) at 2 μg/mL, add increasing concentrations of CTLA-4 x OX40 Bispecific Antibody in 50% Human serum and then add Biotinylated Human CTLA-4, Fc,Avitag (Cat. No. CT4-H82F3) at 0.2 μg/mL. Detection was performed using HRP-conjugated streptavidin with sensitivity of 2 ng/mL (Intact a
Catalog No. 50-210-6734