Learn More
ILEX LIFE SCIENCES LLC HUMAN OXYTOCIN SYNTHETIC 25 MG
SDP
Supplier:  ILEX LIFE SCIENCES LLC A2254025
Encompass_Preferred
Ilex Life Sciences Oxytocin (OT) Human, Synthetic Protein Hormone (25 mg). DESCRIPTION: Our Human Oxytocin (OT) synthetic protein hormone is a single, non-glycosylated, polypeptide chain containing 9 amino acids and has a molecular mass of 1007.2 Dalton. Oxytocin has a molecular formula of C43H66N12O12S2. CAS REGISTRY NO.: 50-56-6. PURITY: Greater than 98.0% as determined by RP-HPLC. AMINO ACID SEQUENCE: H-Cys-Tyr-Ile-Gln-Asn-Cys-Pro-Leu-Gly-NH2. FORMULATION: Sterile filtered white lyophilized powder. The Oxytocin was lyophilized from a concentrated (1 mg/ml) solution with no additives. SYNONYMS: OT, Oxytocin-neurophysin 1, OT-NPI, Ocytocin, Neurophysin 1, α-Hypophamine, Oxytocic hormone, OXT gene product. USAGE: For laboratory research use only; not for use in diagnostic or therapeutic procedures.
Catalog No. 50-273-4009