Learn More
American Research Products Inc Human P2X purinoceptor 7 P2RX
Supplier:  American Research Products Inc CSBEL017325HU
Encompass
Human P2X purinoceptor 7 (P2RX7) ELISA kit from CUSABIO Detection Range 25 pg/mL-1600 pg/mL Sensitivity 6 25 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27482