Learn More
KRISHGEN BIOSYSTEMS Enzyme Immunoassay for the estimation of Programmed Death Ligand-1, PD-L1 / CD274 ELISA in serum and plasma and other biological fluids. Assay Range: 12.5 pg/ml - 600 pg/ml; Sensitivity: 12.5 pg/ml.
Supplier:  KRISHGEN BIOSYSTEMS KBH5925
Encompass_Preferred
Pack Size: 1 x 96 wells. This kit is designed to measure the amount of the PD-L1 bound between a matched antibody pair (sandwich assay). Programmed Cell Death Protein 1 (PD-1) plays a vital role in inhibiting immune responses and promoting self-tolerance through modulating the activity of T-cells, activating apoptosis of antigen-specific T cells and inhibiting apoptosis of regulatory T cells. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Precision CV <10%.
Catalog No. NC2747144