Learn More
Medchemexpress LLC Human PD-L1 inhibitor V TFA | 1485.71 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP2478A5MG
Encompass_Preferred
Human PD-L1 inhibitor V TFA is a human PD-1 protein binding peptide with a Kd value of 3.32 μM. This compound inhibits the interaction of hPD-1/hPD-L1.
  • Inhibits the interaction of hPD-1/hPD-L1
  • Peptide designed with anchor residues Y56, R113, A121, D122, and Y123
  • For research use only
Catalog No. 50-004-23963