Learn More
Sino Biological Human PDCD1 ELISA Pair Set 5PP (Sino Biological)
Supplier:  Sino Biological SEKB103775
Encompass_Preferred
Each vial contains 57 ng of recombinant Human PD-1. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 250 pg/mL is recommended.
Catalog No. 50-237-2323