Learn More
Sino Biological Recombinant Human PDGF-C Protein (His Tag)
Supplier:  Sino Biological 10273H08B20
Encompass_Preferred
A DNA sequence encoding the mature form of human PDGF-C (NP_057289.1) (Val 235-Gly 345) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. NC3588488