Learn More
ACROBiosystems Human PDGF R beta, Fc Tag (PDB-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Phe 530 (Accession # P09619-1).
Supplier:  ACROBiosystems PDBH5259100UG
Encompass_Preferred
Human platelet-derived growth factor receptor, beta polypeptide (PDGFRB) is also called J03278, M21616, CD140B, JTK12, PDGF-R-beta and PDGFR, is receptor that binds specifically to PDGFB and has a tyrosine-protein kinase activity. Is a cell surface tyrosine kinase receptor for members of the platelet-derived growth factor (PDGF) family. The PDGFR/PDGF system includes two receptors (PDGFRA and PDGFRB) and four ligands (A, B, C and D). The receptors PDGFRA and PDGFRB are related in sequence and both are members of the class III subtype of receptor tyrosine kinases (RTKs). Other class III RTKs are CSF1R, KIT and FLT3. PDGF binding induces receptor homo- and heterodimerization and signal transduction. The expression of the α and β receptors is independently regulated in various cell types. Recombinant soluble PDGFRB binds PDGF with high affinity and is potent PDGF antagonist.
Catalog No. 50-223-0962