Learn More
STEMCELL Technologies Human Peripheral Blood CD14+ Monocytes, Frozen, Size: 4 x 10^7 cells
SDP
Supplier:  STEMCELL Technologies 70035
Encompass
Choose ready-to-use, ethically sourced, primary CD14+ monocytes. With personalized service, custom products, flexible delivery times, and the option to reserve entire lots to prescreen cells for applications, we help you get the cells you need. Isolated from mononuclear cells using positive immunomagnetic separation, CD14+ monocytes are collected using Institutional Review Board (IRB)-approved consent forms and protocols and cryopreserved in CryoStor CS10 medium. Use anti-human CD14 antibody clone M5E2 (Catalog #60004AD) for flow cytometry staining. Additional documentation and high-resolution HLA typing (Class I and Class II alleles and CMV status) are available upon request. Acid-citrate-dextrose solution A (ACDA) is added during collection as an anticoagulant. Contact techsupport@stemcell.com for donor specifications (BMI category, smoking status, etc.). Donors are screened for HIV-1, HIV-2, hepatitis B, and hepatitis C. Certain products are only available in select territories.
Catalog No. 50-197-5799