Learn More
STEMCELL Technologies Human Peripheral Blood Leukopak, Fresh, Size: 5 x 10^9 cells minimum
SDP
Supplier:  STEMCELL Technologies 70500.1
Encompass
Obtain large numbers of single-donor mononuclear cells (PBMCs) from normal peripheral blood and reduce the time and reagents required for your downstream cell isolation. Enjoy personalized service, custom products, flexible delivery times, and multiple size options. Leukopaks are highly concentrated, low-volume apheresis collections from normal donors, with Institutional Review Board (IRB)-approved consent forms and protocols. Each full-sized leukopak is produced from ~2 - 3x blood volumes using the Spectra Optia Apheresis System (unless otherwise requested), with ACDA as the anticoagulant. High-resolution HLA typing (Class I and Class II alleles and CMV status) are available upon request. Contact techsupport@stemcell.com for more information, including donor selection or information (BMI category, smoking status, ethnicity, etc.). Donors are recallable and are screened for HIV-1, HIV-2, hepatitis B, and hepatitis C. Certain products are only available in select territories.
Catalog No. NC1129575
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov