Learn More
Sino Biological Human PGM2L1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 12507H02H1
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human PGM2L1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-4269