Learn More
Sino Biological Human pIgR Matched ELISA Antibody Pair Set 5 Plates
Supplier:  Sino Biological SEK101315
Encompass_Preferred
The human PIGR ELISA Pair Set is for the quantitative determinationof human PIGR.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for PIGR coated on a 96-well plate. Standards and samples are added to the wells, and any PIGR present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-PIGR monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of PIGR present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm
Catalog No. 50-007-85533