Learn More
Abcam Human PLCG2 knockout THP-1 cell line
Supplier:  Abcam AB308482
Encompass_Preferred
PLCG2 KO cell line available to order. KO validated by. Free of charge wild type control available. Homozygote. 88 bp deletion. To order both knockout and wild-type control cells: select 2 x 1000000Cells/vial. To order only knockout cells: select 1000000Cells/vial.
Catalog No. NC3839521