Learn More
BIOHIPPO Human PNP Microsample
SDP
Supplier:  BIOHIPPO BHE15212527
Encompass
Human PNP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PNP in serum plasma and other biological fluids It provides a detection range of 1 56-100 ng/mL with a sensitivity of 0 6 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human P NP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human P NP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human P NP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15639