Learn More
Indigo Biosciences Human Peroxisome Proliferator-Activated Receptor delta (PPARd, NR1C2) All-inclusive Cell-Based Luciferase Reporter Assay Kit, 96-well assay plate format
Supplier:  Indigo Biosciences IB00121
Encompass_Preferred
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO’s Human Peroxisome Proliferator-Activated Receptor Delta reporter assay allows you to quickly screen test samples to quantify functional activity, either agonist or antagonist, they may exert against PPAR delta (NR1C2). Kit includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor, optimized media for use during cell thawing and in diluting test samples, reference agonist, Luciferase Detection Reagent, cell culture-ready assay plate, and detailed assay protocol. INDIGO’s unique Cryomite™ process provides exceptional cell viability post-thaw and INDIGO’s strict QA process means consistent results lot to lot (product quality data available upon request). This kit allows researchers to achieve superior sensitivity, clear, reproducible results, and elimina
Catalog No. 50-223-1758