Learn More
Raybiotech Inc Human Pref-1 (DLK-1/FA1) ELISA Kit
Supplier:  Raybiotech Inc ELHPREF11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.4 ng/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human ADAM-12, ADAM-8, B7-H3, BMPR-IB, Cadherin-13, Cadherin-4, CD155, CD229, CD48, CD58, CD84, CD99, CEACAM-5, Coagulation Factor XIV, Cystatin A, Cystatin B, Cystatin E/M, Desmoglein-2, DR3, ErbB4, ESAM, FGF-21, Galectin-2, Galectin-9, I-COS, JAM-A, JAM-B, Kallikrein 5, Midkine, Pentraxin-3, Siglec-10, SLAM, SP-D, Syndecan-4,Testican 2, TIM-3, TLR4, TRAIL, ULBP-1.
Catalog No. 50-194-9659