Learn More
Haematologic Technologies Inc HUMAN PROTEIN S 100UG
Supplier:  Haematologic Technologies Inc HCPS0090
Encompass_Preferred
Coagulation protein, Human Protein S, 100 micrograms, Purifed from plasma, Cofactor, Mode of action: Cofactor for activated protein C, Research use only, 50% (v/v) glycerol/water, store at -20 Celsius.
Catalog No. 50-883-404