Learn More
Sino Biological Recombinant Human PTP1B Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10304H56EB100
Encompass_Preferred
A DNA sequence encoding the Human PTPN1 (NP_002818.1) (Glu2-Asn321) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-3659