Learn More
AMID BIOSCIENCES Amid Biosciences, Human RalB Protein, Recombinant, Biotinylated, 0.1 mg
SDP
Supplier:  AMID BIOSCIENCES RALBB301
Encompass_Preferred
Human RalB is supplied as a recombinant biotinylated small GTPase in a 0.1 mg format. RalB is a member of the Ral branch of the RAS superfamily and is useful for studying downstream signaling effector engagement and small GTPase pathway biology. Applications include SPR, BLI, pull-down, AlphaScreen, RAL-family GTPase signaling studies, GEF interaction assays, small molecule screening. Site-specific N-terminal biotinylation enables direct immobilization on streptavidin surfaces for interaction and screening workflows. Technical specifications are included below.- Quantity - 0.1 mg- Gene - RALB- Mutation / Variant - Wild-type- Organism - Human- Tag - N-terminal 8XHis and AviTag- Expression System - E. coli- Purity - greater than 90% by Coomassie staining- UniProt ID - P11234- Amino Acid Range - 1-206- Buffer - 20 mM Tris-HCl, pH 7.5, 100 mM NaCl, 2 mM DTT, 10% Glycerol- Storage - -80 C- Biotinylated - Yes, N-terminusThis product is for research use only.
Catalog No. NC4043440