Learn More
FUJIFILM BIOSCIENCES INC Human RANTES (CCL5), Recombinant Protein, Shenandoah Biotechnology, 20 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 10013820UG
Encompass_Preferred
Regulated upon activation, normal T cell expressed and secreted (RANTES), also called CCL5, is a chemokine produced by T cells three to five days after T cell activation. RANTES signals through G protein-coupled receptors CCR5, CCR3, CCR1, and through the human CMV-encoded viral receptor US28. RANTES functions to recruit immune cells to inflammatory sites.
Catalog No. 50-900-498