Learn More
BIOHIPPO Human RCBTB2 Microsample
SDP
Supplier:  BIOHIPPO BHE15210965
Encompass
Human RCBTB2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RCBTB2 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 118 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human RCBTB2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human RCBTB2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human RCBTB2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14457