Learn More
STEMCELL Technologies Human Recombinant GM-CSF (E. coli-expressed), Size: 1000 ug
SDP
Supplier:  STEMCELL Technologies 78015.2
Encompass
Use granulocyte-macrophage colony-stimulating factor (GM-CSF) to induce the proliferation and differentiation of hematopoietic progenitor cells and the generation of neutrophils eosinophils and macrophages (Barreda et al. Dev Comp Immunol 2004). Recombinant human GM-CSF also promotes the production of myeloid cells of the granulocytic and monocytic lineages in vivo. It has been tested for mobilization of hematopoietic progenitor cells and for treating chemotherapy-induced neutropenia in patients. In combination with other cytokines such as stem cell factor IL-3 erythropoietin and thrombopoietin GM-CSF also stimulates erythroid and megakaryocyte progenitor cells (Barreda et al.). Human recombinant GM-CSF from STEMCELL comes lyophilized with ≥ 95% purity EC50 200 pg/mL endotoxin levels verified 1.0 EU/μg protein. Alternative names Colony-stimulating factor 2 CSF-2 MGI-1GM Pluripoitin-alpha. Visit STE
Catalog No. 50-239-0135