Learn More
ATCC Human respiratory syncytial virus A strain Long (ATCC VR-26™)
Supplier:  ATCC VR26
Encompass
Human respiratory syncytial virus A strain Long (ATCC VR-26™) is a whole-genome sequenced prototype strain for subgroup A that was isolated in 1956 from a 17-month-old male with pneumonia. This virus is propagated in HEp-2 cells and can be used in respiratory disease research and virucide testing.
Recommended companion products:
HEp-2 cells (Fisher Cat. No. 50-238-5019, ATCC CCL-23™)
EMEM (Fisher Cat. No. 50-238-2632, ATCC 30-2003™)
Fetal bovine serum (Fisher Cat. No. 50-238-3771, ATCC 30-2020™).
Catalog No. 50-238-4232