Learn More
ATCC Human respiratory; syncytial virus; Strain: 18537; RNA
Supplier:  ATCC VR1580D
Encompass
Viral nucleic acids, including viral genomic RNA, in total RNA isolated from Hep-2 cells (ATCC CCL-23) infected with Human respiratory syncytial virus strain 18537 (ATCC VR-1580). Virus and host cells are also available through the ATCC catalog. This product was prepared using methods known to inactivate viruses. RNA isolated from virally infected cells can be used for RT-PCR and other molecular virology applications.
Catalog No. 50-238-2240