Learn More
ATCC Human respiratory; syncytial virus; Strain: 9320
Supplier:  ATCC VR955
Encompass
Human respiratory syncytial virus strain 9320 is propagated in HEp-2 cells (ATCC CCL-23). This strain was isolated in 1977 from the throat swab of a 23-month-old human female with diffuse interstitial pneumonia in Massachusetts. This product in whole-genome sequenced and has applications in respiratory disease research.
Catalog No. 50-238-4215