Learn More
ATCC Human rhinovirus 14; Strain: 1059
Supplier:  ATCC VR284
Encompass
Human rhinovirus 14 strain 1059 is propagated in H1HeLa cells (ATCC CRL-1958). This strain was isolated in 1959 from a throat swab of a young adult human with upper respiratory illness in South Carolina. It has applications in virucide testing and respiratory disease research.
Catalog No. 50-238-2955
 Restricted Item