Learn More
ATCC Human rhinovirus 16; Strain: 11757
Supplier:  ATCC VR283
Encompass
Human rhinovirus 16 strain 11757 is propagated in H1HeLa cells (ATCC CRL-1958). This strain was isolated in 1960 from a throat swab from a healthy 2-year-old human female in Washington, DC. It has applications in respiratory and infectious disease research.
Catalog No. 50-238-1923