Learn More
ATCC Human rhinovirus 17; RNA; Strain: 33342
Supplier:  ATCC VR1663D
Encompass
Genomic RNA isolated from H1 HeLa cells (ATCC CRL-1958) infected with Human rhinovirus 17 strain 33342 (ATCC VR-1663). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. The source organism and host cells are also available through the ATCC catalog.
Catalog No. 50-238-4973